accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1126-61-0

OHCH2HO

CAS RN 1126-61-0

IUPAC Name: 4-(prop-2-en-1-yl)benzene-1,2-diol
Synonyms: 4-allylpyrocatechol
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: FHEHIXJLCWUPCZ-UHFFFAOYSA-N
SMILES: OC1=CC=C(CC=C)C=C1O
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products