accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1127-08-8

OHOOHO

CAS RN 1127-08-8

IUPAC Name: cyclohexane-1,1-dicarboxylic acid
Synonyms: cyclohexane-1,1-dicarboxylic acid
Molecular Weight (g/mol): 172.18
Molecular Formula: C8H12O4
InChi Key: QYQADNCHXSEGJT-UHFFFAOYSA-N
SMILES: OC(=O)C1(CCCCC1)C(O)=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products