CAS RN 112704-79-7
CAS RN 112704-79-7
IUPAC Name:
4-bromo-2-fluorobenzoate
Synonyms:
4-bromo-2-fluorobenzoate
Molecular Weight (g/mol):
218
Molecular Formula:
C7H3BrFO2
InChi Key:
ZQQSRVPOAHYHEL-UHFFFAOYSA-M
SMILES:
[O-]C(=O)C1=CC=C(Br)C=C1F
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar 4-Bromo-2-fluorobenzoic acid, 98%
CAS: 112704-79-7 Molecular Formula: C7H3BrFO2 Molecular Weight (g/mol): 218.00 MDL Number: MFCD00143263 InChI Key: ZQQSRVPOAHYHEL-UHFFFAOYSA-M Synonym: 2-fluoro-4-bromobenzoic acid,4-bromo-2-fluoro-benzoic acid,4-bromo-2-fluorobenzoicacid,benzoic acid, 4-bromo-2-fluoro,pubchem1313,enzalutamide impurity a,acmc-1btmf,2-fluro-4-bromobenzoic acid,ksc174m4p,rarechem al bo 0754 PubChem CID: 302681 IUPAC Name: 4-bromo-2-fluorobenzoic acid SMILES: [O-]C(=O)C1=CC=C(Br)C=C1F