accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 112780-67-3

H3COFMgBr

CAS RN 112780-67-3

IUPAC Name: magnesium(2+) 3-fluoro-4-methoxybenzen-1-ide bromide
Synonyms: magnesium(2+) 3-fluoro-4-methoxybenzen-1-ide bromide
Molecular Weight (g/mol): 229.33
Molecular Formula: C7H6BrFMgO
InChi Key: BUUJBXGCPIVODW-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].COC1=CC=[C-]C=C1F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products