accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1128-08-1

H3CCH3O

CAS RN 1128-08-1

IUPAC Name: 3-methyl-2-pentylcyclopent-2-en-1-one
Synonyms: dihydrojasmone
Molecular Weight (g/mol): 166.26
Molecular Formula: C11H18O
InChi Key: YCIXWYOBMVNGTB-UHFFFAOYSA-N
SMILES: CCCCCC1=C(C)CCC1=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products