CAS RN 112883-40-6
CAS RN 112883-40-6
IUPAC Name:
2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-(methylsulfanyl)butanoic acid
Synonyms:
2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid
Molecular Weight (g/mol):
371.45
Molecular Formula:
C20H21NO4S
InChi Key:
BUBGAUHBELNDEW-UHFFFAOYNA-N
SMILES:
CSCCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
1
–
1
of
1
results
1
N-Fmoc-D-methionine, 98%, Thermo Scientific™
CAS: 112883-40-6 Molecular Formula: C20H21NO4S Molecular Weight (g/mol): 371.45 MDL Number: MFCD00062958 MFCD00037134 MFCD00062958 InChI Key: BUBGAUHBELNDEW-GOSISDBHSA-N Synonym: fmoc-d-met-oh,fmoc-d-methionine,n-9h-fluoren-9-ylmethoxy carbonyl-d-methionine,r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-methylthio butanoic acid,d-methionine, n-9h-fluoren-9-ylmethoxy carbonyl,2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methylsulfanyl butanoic acid,n-fmoc-d-methionine,ambotzfaa1145,n-9-fluorenylmethyloxycarbonyl-d-methionine PubChem CID: 6992521 IUPAC Name: (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylsulfanylbutanoic acid SMILES: CSCC[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O