CAS RN 112965-21-6
CAS RN 112965-21-6
IUPAC Name:
(1R,3S,5Z)-5-{2-[(1R,4E,7aR)-1-[(2R,3E,5S)-5-cyclopropyl-5-hydroxypent-3-en-2-yl]-7a-methyl-octahydro-1H-inden-4-ylidene]ethylidene}-4-methylidenecyclohexane-1,3-diol
Synonyms:
calcipotriol
Molecular Weight (g/mol):
412.61
Molecular Formula:
C27H40O3
InChi Key:
LWQQLNNNIPYSNX-OUSHRDKSSA-N
SMILES:
C[C@H](\C=C\[C@@H](O)C1CC1)[C@H]1CCC2\C(CCC[C@]12C)=C\C=C1\C[C@@H](O)C[C@H](O)C1=C
1
–
2
of
2
results
1
Tocris Bioscience™ Calcipotriol

CAS: 112965-21-6 Molecular Formula: C27H40O3 Molecular Weight (g/mol): 412.61
2
Medchem Express Calcipotriol, MedChemExpress
MedChemExpress Calcipotriol is a synthetic VitD3 analogue with a high affinity for the vitamin D receptor.
