CAS RN 1129669-05-1
CAS RN 1129669-05-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express IM-12, MedChemExpress
MedChemExpress IM-12 is an inhibitor of GSK-3β, with an IC50 of 53 nM, and also enhances Wnt signalling.
