accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 112981-69-8

FFFFFFMgBr

CAS RN 112981-69-8

IUPAC Name: magnesium(2+) 3,5-bis(trifluoromethyl)benzen-1-ide bromide
Synonyms: magnesium(2+) 3,5-bis(trifluoromethyl)benzen-1-ide bromide
Molecular Weight (g/mol): 317.31
Molecular Formula: C8H3BrF6Mg
InChi Key: SHAKXLPGFQFNJG-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].FC(F)(F)C1=CC(=C[C-]=C1)C(F)(F)F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products