accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 113-48-4

H3CH3CNOO

CAS RN 113-48-4

IUPAC Name: 4-(2-ethylhexyl)-4-azatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
Synonyms: ethylhexylbicycloheptene dicarboximide
Molecular Weight (g/mol): 275.39
Molecular Formula: C17H25NO2
InChi Key: WLLGXSLBOPFWQV-UHFFFAOYNA-N
SMILES: CCCCC(CC)CN1C(=O)C2C3CC(C=C3)C2C1=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products