accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 113-92-8

H3CNHCH3ClNOHOOO(Z)

CAS RN 113-92-8

IUPAC Name: [3-(4-chlorophenyl)-3-(pyridin-2-yl)propyl]dimethylazanium (2Z)-3-carboxyprop-2-enoate
Synonyms: chlorphenamine
Molecular Weight (g/mol): 390.86
Molecular Formula: C20H23ClN2O4
InChi Key: DBAKFASWICGISY-BTJKTKAUNA-N
SMILES: OC(=O)\C=C/C([O-])=O.C[NH+](C)CCC(C1=CC=C(Cl)C=C1)C1=CC=CC=N1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products