accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 113023-73-7

H3COHOBr

CAS RN 113023-73-7

IUPAC Name: 4-(1-bromoethyl)benzoic acid
Synonyms: 4-(1-bromoethyl)benzoic acid
Molecular Weight (g/mol): 229.07
Molecular Formula: C9H9BrO2
InChi Key: VICCYULHZWEWMB-UHFFFAOYNA-N
SMILES: CC(Br)C1=CC=C(C=C1)C(O)=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products