accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1135695-98-5

CAS RN 1135695-98-5

IUPAC Name: 5-(2,6-difluorophenoxy)-3-{3-methyl-2-[(quinolin-2-yl)formamido]butanamido}-4-oxopentanoic acid
Synonyms: 5-(2,6-difluorophenoxy)-3-[3-methyl-2-(quinolin-2-ylformamido)butanamido]-4-oxopentanoic acid
Molecular Weight (g/mol): 513.5
Molecular Formula: C26H25F2N3O6
InChi Key: OOBJCYKITXPCNS-UHFFFAOYNA-N
SMILES: CC(C)C(NC(=O)C1=CC=C2C=CC=CC2=N1)C(=O)NC(CC(O)=O)C(=O)COC1=C(F)C=CC=C1F
Grade 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products