accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1136466-93-7

NHNOOOH

CAS RN 1136466-93-7

IUPAC Name: 6-(N'-cyclopentylidenehydrazinecarbonyl)hexanoic acid
Synonyms: 6-(N'-cyclopentylidenehydrazinecarbonyl)hexanoic acid
Molecular Weight (g/mol): 240.3
Molecular Formula: C12H20N2O3
InChi Key: CVYPYYPTFNVREL-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCC(=O)NN=C1CCCC1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products