accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1137-73-1

HOONOHO

CAS RN 1137-73-1

IUPAC Name: 2-[(carboxymethyl)(phenyl)amino]acetic acid
Synonyms: [(carboxymethyl)(phenyl)amino]acetic acid
Molecular Weight (g/mol): 209.2
Molecular Formula: C10H11NO4
InChi Key: GQBWTAGIANQVGB-UHFFFAOYSA-N
SMILES: OC(=O)CN(CC(O)=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products