accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 114-83-0

H3COHNNH

CAS RN 114-83-0

IUPAC Name: N'-phenylacetohydrazide
Synonyms: N(1)-acetylphenylhydrazine
Molecular Weight (g/mol): 150.18
Molecular Formula: C8H10N2O
InChi Key: UICBCXONCUFSOI-UHFFFAOYSA-N
SMILES: CC(=O)NNC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products