CAS RN 1140525-25-2
CAS RN 1140525-25-2
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
Tocris Bioscience™ SB 290157 trifluoroacetate

Potent C3a receptor antagonist
2
SB290157 trifluoroacetate, MedChemExpress
MedChemExpress SB290157 trifluoroacetate is a potent and selective C3a receptor antagonist with an IC50 of 200 nM.
