accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1142363-56-1

BOOH3CCH3H3CH3COCH3H3CCH3CH3

CAS RN 1142363-56-1

IUPAC Name: 4,4,5,5-tetramethyl-2-(2,2,6,6-tetramethyl-3,6-dihydro-2H-pyran-4-yl)-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-(2,2,6,6-tetramethyl-3H-pyran-4-yl)-1,3,2-dioxaborolane
Molecular Weight (g/mol): 266.19
Molecular Formula: C15H27BO3
InChi Key: VOXXPMAJNSVUKJ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(C)(C)OC(C)(C)C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products