CAS RN 114289-47-3
CAS RN 114289-47-3
IUPAC Name:
2,2-dimethyl-N-(2,4,6-trimethoxyphenyl)dodecanamide
Synonyms:
2,2-dimethyl-N-(2,4,6-trimethoxyphenyl)dodecanamide
Molecular Weight (g/mol):
393.57
Molecular Formula:
C23H39NO4
InChi Key:
WAFNZAURAWBNDZ-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCC(C)(C)C(=O)NC1=C(OC)C=C(OC)C=C1OC
1
–
1
of
1
results
1
Tocris CI 976, Tocris Bioscienceâ„¢
CAS: 114289-47-3 Molecular Formula: C23H39NO4 Molecular Weight (g/mol): 393.568 InChI Key: WAFNZAURAWBNDZ-UHFFFAOYSA-N Synonym: 2,2-dimethyl-n-2,4,6-trimethoxyphenyl dodecanamide,chembl22373,unii-8653iq20ea,dodecanamide,2,2-dimethyl-n-2,4,6-trimethoxyphenyl,dodecanamide, 2,2-dimethyl-n-2,4,6-trimethoxyphenyl,2,2-dimethyl-dodecanoic acid 2,4,6-trimethoxy-phenyl-amide,acmc-1cuxf,d0d1xg,ci hplc , solid PubChem CID: 122327 IUPAC Name: 2,2-dimethyl-N-(2,4,6-trimethoxyphenyl)dodecanamide SMILES: CCCCCCCCCCC(C)(C)C(=O)NC1=C(C=C(C=C1OC)OC)OC