accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 114369-43-6

ClNNNN

CAS RN 114369-43-6

IUPAC Name: 4-(4-chlorophenyl)-2-phenyl-2-[(1H-1,2,4-triazol-1-yl)methyl]butanenitrile
Synonyms: fenbuconazole 44CLPHENYL2PH21H124TRIAZOL1ylmethylbutyronitrile
Molecular Weight (g/mol): 336.82
Molecular Formula: C19H17ClN4
InChi Key: RQDJADAKIFFEKQ-UHFFFAOYNA-N
SMILES: ClC1=CC=C(CCC(CN2C=NC=N2)(C#N)C2=CC=CC=C2)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Concentration 
  • (2)
  • (2)
Solvent or Matrix 
  • (2)
  • (2)
Grade 
  • (1)
Industry Type 
  • (1)

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products