CAS RN 114435-02-8
CAS RN 114435-02-8
IUPAC Name:
4-fluoro-1,3-dioxolan-2-one
Synonyms:
4-fluoro-1,3-dioxolan-2-one
Molecular Weight (g/mol):
106.05
Molecular Formula:
C3H3FO3
InChi Key:
SBLRHMKNNHXPHG-UHFFFAOYNA-N
SMILES:
FC1COC(=O)O1
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific Alfa Aesar 4-Fluoro-1,3-dioxolan-2-one, 98%
Thermo Scientific Alfa Aesar 4-Fluoro-1,3-dioxolan-2-one, 98%
CAS: 114435-02-8 Molecular Formula: C3H3FO3 Molecular Weight (g/mol): 106.052 MDL Number: MFCD06247543 InChI Key: SBLRHMKNNHXPHG-UHFFFAOYSA-N Synonym: fluoroethylene carbonate,1,3-dioxolan-2-one, 4-fluoro,fluoro ethylene carbonate,4-fluoro-1,3-dioxalan-2-one,4-fluoro-2-oxo-1,3-dioxolane,4-fluoro-ethylene carbonate,4-fluoro-ethylene carboante,ksc219e1h,fec, lithium ion battery grade PubChem CID: 2769656 IUPAC Name: 4-fluoro-1,3-dioxolan-2-one SMILES: C1C(OC(=O)O1)F