accessibility menu, dialog, popup

Test

CAS RN 1147-23-5

CAS RN 1147-23-5

IUPAC Name: 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodo-1,2-dihydropyrimidin-2-one
Synonyms: 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidin-2-one
Molecular Weight (g/mol): 369.12
Molecular Formula: C9H12IN3O5
InChi Key: LQQGJDJXUSAEMZ-UAKXSSHOSA-N
SMILES: NC1=NC(=O)N(C=C1I)[C@@H]1O[C@H](CO)[C@@H](O)[C@H]1O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results