accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 114745-26-5

CH3CH3ONHNONHNN(R)(R)(R)(R)OOHOOH3COH3COH3C

CAS RN 114745-26-5

IUPAC Name: N-[9-(5-{[bis(4-methoxyphenyl)(phenyl)methoxy]methyl}-4-hydroxy-3-methoxyoxolan-2-yl)-6-oxo-6,9-dihydro-3H-purin-2-yl]-2-methylpropanamide
Synonyms: N-[9-(5-{[bis(4-methoxyphenyl)(phenyl)methoxy]methyl}-4-hydroxy-3-methoxyoxolan-2-yl)-6-oxo-3H-purin-2-yl]-2-methylpropanamide
Molecular Weight (g/mol): 669.74
Molecular Formula: C36H39N5O8
InChi Key: ISQLJOGRNUQHJX-UHFFFAOYNA-N
SMILES: COC1C(O)C(COC(C2=CC=CC=C2)(C2=CC=C(OC)C=C2)C2=CC=C(OC)C=C2)OC1N1C=NC2=C1NC(NC(=O)C(C)C)=NC2=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products