accessibility menu, dialog, popup

Test

CAS RN 114772-40-6

CAS RN 114772-40-6

IUPAC Name: tert-butyl 4'-(bromomethyl)-[1,1'-biphenyl]-2-carboxylate
Synonyms: tert-butyl 4'-(bromomethyl)-[1,1'-biphenyl]-2-carboxylate
Molecular Weight (g/mol): 347.25
Molecular Formula: C18H19BrO2
InChi Key: YHXCWNQNVMAENQ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)C1=CC=CC=C1C1=CC=C(CBr)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results