accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 114787-91-6

CH3HOOCH3

CAS RN 114787-91-6

IUPAC Name: (4-methoxy-3-methylphenyl)methanol
Synonyms: (4-methoxy-3-methylphenyl)methanol
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: UVIIQYUUEVQCOR-UHFFFAOYSA-N
SMILES: COC1=C(C)C=C(CO)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products