accessibility menu, dialog, popup

UserName

The Ultimate Lab Appliances Promotion

Save Up to 78%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 114873-03-9

H3CH3CCH3OONH(S)ClOHO

CAS RN 114873-03-9

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(3-chlorophenyl)propanoic acid
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-3-(3-chlorophenyl)propanoic acid
Molecular Weight (g/mol): 299.75
Molecular Formula: C14H18ClNO4
InChi Key: RCZHBTHQISEPPP-NSHDSACASA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](CC1=CC=CC(Cl)=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products