CAS RN 114926-37-3
CAS RN 114926-37-3
IUPAC Name:
(2S)-2-amino-3-(3-methylphenyl)propanoic acid
Synonyms:
3-methylphenylalanine
Molecular Weight (g/mol):
179.22
Molecular Formula:
C10H13NO2
InChi Key:
JZRBSTONIYRNRI-VIFPVBQESA-N
SMILES:
CC1=CC=CC(C[C@H](N)C(O)=O)=C1
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar 3-Methyl-L-phenylalanine, 95%
CAS: 114926-37-3 Molecular Formula: C10H13NO2 Molecular Weight (g/mol): 179.219 MDL Number: MFCD00044972 InChI Key: JZRBSTONIYRNRI-VIFPVBQESA-N Synonym: 3-methyl-l-phenylalanine,l-3-methylphenylalanine,3-methylphenyl-l-alanine,3-methy-l-phenylalanine,s-2-amino-3-m-tolyl propanoic acid,3-methylphenylalanine,l-3-methylphe,phenylalanine, 3-methyl,2s-2-amino-3-3-methylphenyl propanoic acid,l-3-me-phe-oh PubChem CID: 2761494 IUPAC Name: (2S)-2-amino-3-(3-methylphenyl)propanoic acid SMILES: CC1=CC(=CC=C1)CC(C(=O)O)N