accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 115-32-2

HOClClClClCl

CAS RN 115-32-2

IUPAC Name: 2,2,2-trichloro-1,1-bis(4-chlorophenyl)ethan-1-ol
Synonyms: dicofol caswell no. 093 DTMC mitigan keltane para,para'-kelthane kelthane dust base C14H9cl5O kelthane cekudifol
Molecular Weight (g/mol): 370.48
Molecular Formula: C14H9Cl5O
InChi Key: UOAMTSKGCBMZTC-UHFFFAOYSA-N
SMILES: OC(C1=CC=C(Cl)C=C1)(C1=CC=C(Cl)C=C1)C(Cl)(Cl)Cl
Molecular Weight (g/mol) 
  • (7)
  • (1)
Solvent or Matrix 
  • (2)
  • (2)
  • (2)
  • (1)
Concentration 
  • (3)
  • (3)

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (1)
  • (1)
  • (7)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products