CAS RN 115-33-3
CAS RN 115-33-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express Oxyphenisatin acetate, MedChemExpress
MedChemExpress Oxyphenisatin acetate, the pro-drug of oxyphenisatin, is used to be a laxative.
