accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1150-62-5

N

CAS RN 1150-62-5

IUPAC Name: 9-phenyl-9H-carbazole
Synonyms: 9-phenylcarbazole
Molecular Weight (g/mol): 243.31
Molecular Formula: C18H13N
InChi Key: VIJYEGDOKCKUOL-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products