accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1152197-23-3

CAS RN 1152197-23-3

IUPAC Name: [(6-{6-amino-8-[(4-chlorophenyl)sulfanyl]-9H-purin-9-yl}-7-methoxy-2-oxo-hexahydro-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-2-yl)oxy]methyl acetate
Synonyms: [(6-{6-amino-8-[(4-chlorophenyl)sulfanyl]purin-9-yl}-7-methoxy-2-oxo-tetrahydro-4H-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-2-yl)oxy]methyl acetate
Molecular Weight (g/mol): 557.9
Molecular Formula: C20H21ClN5O8PS
InChi Key: FZMWUFYPEVDWPA-UHFFFAOYNA-N
SMILES: COC1C(OC2COP(=O)(OCOC(C)=O)OC12)N1C(SC2=CC=C(Cl)C=C2)=NC2=C(N)N=CN=C12

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products