accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 115297-57-9

H2NNH2Cl

CAS RN 115297-57-9

IUPAC Name: [amino(4-chlorophenyl)methylidene]azanium
Synonyms: [amino(4-chlorophenyl)methylidene]azanium
Molecular Weight (g/mol): 155.6
Molecular Formula: C7H8ClN2
InChi Key: LFCUMSZYMJRUHD-UHFFFAOYSA-O
SMILES: NC(=[NH2+])C1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products