accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 115314-17-5

ONOSOOO(R)O

CAS RN 115314-17-5

IUPAC Name: [(2R)-oxiran-2-yl]methyl 3-nitrobenzene-1-sulfonate
Synonyms: (2R)-oxiran-2-ylmethyl 3-nitrobenzenesulfonate
Molecular Weight (g/mol): 259.23
Molecular Formula: C9H9NO6S
InChi Key: AIHIHVZYAAMDPM-MRVPVSSYSA-N
SMILES: [O-][N+](=O)C1=CC=CC(=C1)S(=O)(=O)OC[C@H]1CO1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products