accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1154-25-2

CH3NCH3CH3NNH2NClONNH2H2N

CAS RN 1154-25-2

IUPAC Name: 3-amino-6-chloro-N-(diaminomethylidene)-5-[ethyl(propan-2-yl)amino]pyrazine-2-carboxamide
Synonyms: 3-amino-6-chloro-N-(diaminomethylidene)-5-[ethyl(isopropyl)amino]pyrazine-2-carboxamide
Molecular Weight (g/mol): 299.76
Molecular Formula: C11H18ClN7O
InChi Key: QDERNBXNXJCIQK-UHFFFAOYSA-N
SMILES: CCN(C(C)C)C1=NC(N)=C(N=C1Cl)C(=O)N=C(N)N
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Grade 
  • (2)

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products