accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1155-62-0

HOONHOOOHO

CAS RN 1155-62-0

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}pentanedioic acid
Synonyms: 2-{[(benzyloxy)carbonyl]amino}pentanedioic acid
Molecular Weight (g/mol): 281.26
Molecular Formula: C13H15NO6
InChi Key: PVFCXMDXBIEMQG-UHFFFAOYNA-N
SMILES: OC(=O)CCC(NC(=O)OCC1=CC=CC=C1)C(O)=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products