CAS RN 115685-56-8
CAS RN 115685-56-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar Lanthanum(III) 2-methoxyethoxide, 99.9% (REO), 5% w/v in 2-methoxyethanol
CAS: 115685-56-8 MDL Number: MFCD00210619 Synonym: lanthanum iii 2-methoxyethoxide,lanthanum tris 1-methoxyethan-1-olate,acmc-20akoi,lanthanum 3+ ion tris 1-methoxyethanolate