accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1159-53-1

CH3NH3CCH3

CAS RN 1159-53-1

IUPAC Name: 4-methyl-N,N-bis(4-methylphenyl)aniline
Synonyms: 4-methyl-N,N-bis(4-methylphenyl)aniline
Molecular Weight (g/mol): 287.41
Molecular Formula: C21H21N
InChi Key: YXYUIABODWXVIK-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)N(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products