CAS RN 1159824-67-5
CAS RN 1159824-67-5
IUPAC Name:
5-{2-amino-8-fluoro-[1,2,4]triazolo[1,5-a]pyridin-6-yl}-N-tert-butylpyridine-3-sulfonamide
Synonyms:
5-{2-amino-8-fluoro-[1,2,4]triazolo[1,5-a]pyridin-6-yl}-N-tert-butylpyridine-3-sulfonamide
Molecular Weight (g/mol):
364.4
Molecular Formula:
C15H17FN6O2S
InChi Key:
RXRZPHQBTHQXSV-UHFFFAOYSA-N
SMILES:
CC(C)(C)NS(=O)(=O)C1=CN=CC(=C1)C1=CN2N=C(N)N=C2C(F)=C1
1
–
1
of
1
results
1
CZC24832, MedChemExpress
MedChemExpress CZC24832 is a highly selective and potent PI3Kγ inhibitor (IC50=27 nM) with apparent dissociation constants (Kdapp) of 19 nM.
