accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1160927-48-9

NHNOOOHOHO(E/Z)

CAS RN 1160927-48-9

IUPAC Name: 2-{[(6-carboxyhexanamido)imino]methyl}benzoic acid
Synonyms: 2-{[(6-carboxyhexanamido)imino]methyl}benzoic acid
Molecular Weight (g/mol): 306.32
Molecular Formula: C15H18N2O5
InChi Key: ABKJCDILEUEJSH-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCC(=O)NN=CC1=CC=CC=C1C(O)=O
Color 
  • (5)
Grade 
  • (5)

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (7)
  • (2)
  • (5)
  • (5)
  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products