accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1165-91-9

ClClOOOOClClClCl

CAS RN 1165-91-9

IUPAC Name: bis(2,4,6-trichlorophenyl) oxalate
Synonyms: TCPO
Molecular Weight (g/mol): 448.88
Molecular Formula: C14H4Cl6O4
InChi Key: GEVPIWPYWJZSPR-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C(OC(=O)C(=O)OC2=C(Cl)C=C(Cl)C=C2Cl)C(Cl)=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products