accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 116686-15-8

CH3ONHSH3COOOFF

CAS RN 116686-15-8

IUPAC Name: N-[4-acetyl-2-(2,4-difluorophenoxy)phenyl]methanesulfonamide
Synonyms: N-[4-acetyl-2-(2,4-difluorophenoxy)phenyl]methanesulfonamide
Molecular Weight (g/mol): 341.33
Molecular Formula: C15H13F2NO4S
InChi Key: DIIYLGZNZGPXRR-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(NS(C)(=O)=O)C(OC2=CC=C(F)C=C2F)=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products