accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 117-82-8

H3COOOOOOCH3

CAS RN 117-82-8

IUPAC Name: 1,2-bis(2-methoxyethyl) benzene-1,2-dicarboxylate
Synonyms: bis(methoxyethyl)phthalate
Molecular Weight (g/mol): 282.29
Molecular Formula: C14H18O6
InChi Key: HSUIVCLOAAJSRE-UHFFFAOYSA-N
SMILES: COCCOC(=O)C1=CC=CC=C1C(=O)OCCOC

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products