accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1170613-55-4

CH3NOHFH3C(E/Z)(E/Z)

CAS RN 1170613-55-4

IUPAC Name: N-[1-(4-fluorophenyl)-2-methylpent-1-en-3-ylidene]hydroxylamine
Synonyms: N-[1-(4-fluorophenyl)-2-methylpent-1-en-3-ylidene]hydroxylamine
Molecular Weight (g/mol): 207.25
Molecular Formula: C12H14FNO
InChi Key: HKROEBDHHKMNBZ-UHFFFAOYSA-N
SMILES: CCC(=NO)C(C)=CC1=CC=C(F)C=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (5)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products