accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 117106-20-4

H3C(R)O(S)NOHOH3CCH3CH3OOH3C

CAS RN 117106-20-4

IUPAC Name: 3-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)butanoic acid
Synonyms: 3-(tert-butoxy)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl](methyl)amino}butanoic acid
Molecular Weight (g/mol): 411.5
Molecular Formula: C24H29NO5
InChi Key: VIUVLZHFMIFLHU-UHFFFAOYNA-N
SMILES: CC(OC(C)(C)C)C(N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products