CAS RN 1172133-28-6
CAS RN 1172133-28-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express HO-3867, MedChemExpress
MedChemExpress HO-3867 is a selective and potent STAT3 inhibitor and shows good antitumor activity.
