accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1174064-61-9

CH3NHNNH2H3CCH3H3C

CAS RN 1174064-61-9

IUPAC Name: 5,5,7,7-tetramethyl-1H,5H,6H,7H-cyclopenta[f]indazol-4-amine
Synonyms: 5,5,7,7-tetramethyl-1H,6H-cyclopenta[f]indazol-4-amine
Molecular Weight (g/mol): 229.33
Molecular Formula: C14H19N3
InChi Key: IOJZKUQKQPKIQK-UHFFFAOYSA-N
SMILES: CC1(C)CC(C)(C)C2=C1C=C1NN=CC1=C2N

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products