accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1174157-65-3

CAS RN 1174157-65-3

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 3-(prop-2-yn-1-yloxy)propanoate
Synonyms: 2,5-dioxopyrrolidin-1-yl 3-(prop-2-yn-1-yloxy)propanoate
Molecular Weight (g/mol): 225.2
Molecular Formula: C10H11NO5
InChi Key: WKIKHHMUNOVQLD-UHFFFAOYSA-N
SMILES: O=C(CCOCC#C)ON1C(=O)CCC1=O
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products