accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 117500-61-5

CH3NH3COClHCl

CAS RN 117500-61-5

IUPAC Name: 3-(dimethylamino)benzoyl chloride hydrochloride
Synonyms: 3-(dimethylamino)benzoyl chloride hydrochloride
Molecular Weight (g/mol): 220.09
Molecular Formula: C9H11Cl2NO
InChi Key: NUDQPATXNFWFNK-UHFFFAOYSA-N
SMILES: Cl.CN(C)C1=CC=CC(=C1)C(Cl)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products