accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 117572-79-9

H3COBrN

CAS RN 117572-79-9

IUPAC Name: 3-bromo-4-methoxybenzonitrile
Synonyms: 3-bromo-4-methoxybenzonitrile
Molecular Weight (g/mol): 212.05
Molecular Formula: C8H6BrNO
InChi Key: QHWZMDRKTYTPEE-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1Br)C#N
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products