CAS RN 117690-79-6
CAS RN 117690-79-6
IUPAC Name:
1-(5-ethyl-2-hydroxy-4-{[6-methyl-6-(2H-1,2,3,4-tetrazol-5-yl)heptyl]oxy}phenyl)ethan-1-one
Synonyms:
1-(5-ethyl-2-hydroxy-4-{[6-methyl-6-(2H-1,2,3,4-tetrazol-5-yl)heptyl]oxy}phenyl)ethanone
Molecular Weight (g/mol):
360.46
Molecular Formula:
C19H28N4O3
InChi Key:
WCGXJPFHTHQNJL-UHFFFAOYSA-N
SMILES:
CCC1=CC(C(C)=O)=C(O)C=C1OCCCCCC(C)(C)C1=NNN=N1
1
–
1
of
1
results
1
Tocris Bioscience™ LY 255283

CAS: 117690-79-6 Molecular Formula: C19H28N4O3 Molecular Weight (g/mol): 360.458